2-Fluoro-α-methylbenzyl alcohol 98%

Supplier: Ambeed
Danger

A208898-250MG A208898-5G A208898-1G
77192-562EA 6 USD
77192-562 77192-564 77568-002
2-Fluoro-α-methylbenzyl alcohol 98%
2-Fluoro-α-methylbenzyl alcohol

Precaución: For research use only.

Formula: C₈H₉FO
MW: 140.16
MDL Number: MFCD00014401
CAS Number: 445-26-1
UN: 1993
ADR: 3 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR