1-(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)cyclopropanecarbonitrile 97%

Supplier: Ambeed
Warning

A208137-1G A208137-5G
77164-884EA 15 USD
77164-884 77164-886
1-(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)cyclopropanecarbonitrile 97%
1-(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)cyclopropanecarbonitrile

Precaución: for Research Use Only

Formula: C₁₁H₇F₂NO₂
MDL Number: MFCD14155913
CAS Number: 862574-87-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR