4-Aminobutyraldehyde diethyl acetal 97%

Supplier: Ambeed
Danger

Synonyms: 4,4-Diethoxybutan-1-amine, 4-Aminobutanal diethyl acetal

A207988-100G A207988-10G A207988-1G A207988-25G A207988-5G
77192-552EA 159.65 USD
77192-552 77192-554 77192-556 77192-558 77192-560
4-Aminobutyraldehyde diethyl acetal 97%
4-Aminobutyraldehyde diethyl acetal

Precaución: for Research Use Only

Formula: C₈H₁₉NO₂
MW: 161.24 g/mol
MDL Number: MFCD00008227
CAS Number: 6346-09-4
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR