4,7-Dibromobenzo[c][1,2,5]thiadiazole-5,6-diamine 95%

Supplier: Ambeed
Warning

A207868-1G A207868-250MG A207868-5G A207868-100MG
77164-870EA 128.75 USD
77164-870 77164-872 77550-334 77683-590
4,7-Dibromobenzo[c][1,2,5]thiadiazole-5,6-diamine 95%
4,7-Dibromobenzo[c][1,2,5]thiadiazole-5,6-diamine

Precaución: For research use only.

Formula: C₆H₄Br₂N₄S
MW: 323.99 g/mol
MDL Number: MFCD23136883
CAS Number: 141215-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR