2-Amino-5-chloropyrazine 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A207833-1G A207833-25G A207833-10G A207833-5G
77164-860EA 9 USD
77164-860 77164-862 77683-589 77892-306
2-Amino-5-chloropyrazine 98%, Ambeed.Inc
2-Amino-5-chloropyrazine

Precaución: For research use only.

Formula: C₄H₄ClN₃
MDL Number: MFCD03423596
CAS Number: 33332-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR