Citarinostat 98+%

Supplier: Ambeed
Warning

Synonyms: 2-((2-chlorophenyl)(phenyl)amino)-N-(7-(hydroxyamino)-7-oxoheptyl)pyrimidine-5-carboxamide, ACY-241

A207811-1MG A207811-5MG
77164-856EA 39.14 USD
77164-856 77164-858
Citarinostat 98+%
Citarinostat

Precaución: for Research Use Only

Formula: C₂₄H₂₆ClN₅O₃
MW: 467.95 g/mol
MDL Number: MFCD28023593
CAS Number: 1316215-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR