3-Methoxybenzamidine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A207732-10G
A207732-1G
A207732-25G
A207732-5G
77164-852EA
36
USD
77164-852
77164-854
77892-304
77892-305
3-Methoxybenzamidine hydrochloride 95%
3-Methoxybenzamidine hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₁ClN₂O |
MDL Number:
MFCD04114430 CAS Number: 26113-44-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...