3-((4-(Trifluoromethyl)phenyl)amino)benzoic acid 95%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: AKR1C3-IN-4

A207518-100MG A207518-250MG A207518-1G A207518-5G
77164-836EA 91.67 USD
77164-836 77164-838 77683-586 77683-587
3-((4-(Trifluoromethyl)phenyl)amino)benzoic acid 95%, Ambeed.Inc
3-((4-(Trifluoromethyl)phenyl)amino)benzoic acid

Precaución: For research use only.

Formula: C₁₄H₁₀F₃NO₂
MW: 281.23 g/mol
MDL Number: MFCD28016823
CAS Number: 1284180-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR