1-(4-Aminobutyl)guanidine dihydrochloride 95%

Supplier: Ambeed
Warning

A207474-1G A207474-250MG
77192-532EA 22 USD
77192-532 77192-534
1-(4-Aminobutyl)guanidine dihydrochloride 95%
1-(4-Aminobutyl)guanidine dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₆Cl₂N₄
MW: 203.11 g/mol
MDL Number: MFCD11053502
CAS Number: 334-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR