(S)-4-Isopropyl-5,5-dimethyloxazolidin-2-one 97%
Supplier: Ambeed
|
Warning
|
A207419-100MG
A207419-250MG
A207419-1G
77192-528EA
39.14
USD
77192-528
77192-530
77593-352
(S)-4-Isopropyl-5,5-dimethyloxazolidin-2-one 97%
(S)-4-Isopropyl-5,5-dimethyloxazolidin-2-one
Precaución: for Research Use Only
Formula:
C₈H₁₅NO₂ MW: 157.21 g/mol |
MDL Number:
MFCD01321231 CAS Number: 168297-86-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...