1,2-Phenylenediacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: o-Phenylenediacetic acid, 2,2'-(1,2-Phenylene)diacetic acid

A207319-10G A207319-1G A207319-5G A207319-25G A207319-100G
77192-508EA 24 USD
77192-508 77192-510 77192-512 77615-742 77615-740
1,2-Phenylenediacetic acid 98%
1,2-Phenylenediacetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
MDL Number: MFCD00004329
CAS Number: 7500-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR