6-Aminopyridin-3-ol 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Aminopyridin-3-ol, 2-Amino-5-hydroxypyridine
A207270-100MG
A207270-10G
A207270-1G
A207270-250MG
A207270-5G
77164-822EA
13
USD
77164-822
77164-824
77164-826
77164-828
77164-830
6-Aminopyridin-3-ol 95%
6-Aminopyridin-3-ol
Precaución: for Research Use Only
Formula:
C₅H₆N₂O MW: 110.1100 g/mol |
MDL Number:
MFCD03541056 CAS Number: 55717-46-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...