5-Chloroisatoic anhydride 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-1H-benzo[d][1,3]oxazine-2,4-dione

A206502-100G A206502-25G A206502-5G
77192-494EA 128.75 USD
77192-494 77192-496 77192-498
5-Chloroisatoic anhydride 97%
5-Chloroisatoic anhydride

Precaución: for Research Use Only

Formula: C₈H₄ClNO₃
MW: 197.58 g/mol
MDL Number: MFCD00006701
CAS Number: 4743-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR