Thiazol-2-ylmethanamine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Aminomethyl)-1,3-thiazole hydrochloride, (1,3-Thiazol-2-yl)methylamine hydrochloride, 2-Aminomethylthiazole hydrochloride

A206373-1G A206373-250MG A206373-5G
77164-788EA 47 USD
77164-788 77164-790 77892-297
Thiazol-2-ylmethanamine hydrochloride 98%
Thiazol-2-ylmethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₄H₇ClN₂S
MW: 150.63 g/mol
MDL Number: MFCD04113607
CAS Number: 850852-85-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR