Methyl propionylacetate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-oxovalerate, Methyl 3-oxo-pentanoate

A206153-1000G A206153-100G A206153-1G A206153-25G A206153-500G A206153-5G
77164-774EA 141.11 USD
77164-774 77164-776 77164-778 77164-780 77164-782 77164-784
Methyl propionylacetate 98%
Methyl propionylacetate

Precaución: for Research Use Only

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
MDL Number: MFCD00011705
CAS Number: 30414-53-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR