7-Fluorobenzo[d]thiazole 97%

Supplier: Ambeed
Warning

A206039-100MG A206039-1G A206039-250MG A206039-50MG
77192-480EA 15 USD
77192-480 77192-482 77192-484 77192-486
7-Fluorobenzo[d]thiazole 97%
7-Fluorobenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₇H₄FNS
CAS Number: 214855-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR