6-Methylpiperidin-2-one 95%
Supplier: Ambeed
|
Warning
|
A205841-1G
A205841-5G
A205841-250MG
A205841-25G
77164-762EA
35
USD
77164-762
77164-764
77892-293
77892-294
6-Methylpiperidin-2-one 95%
6-Methylpiperidin-2-one
Precaución: for Research Use Only
Formula:
C₆H₁₁NO |
MDL Number:
MFCD00127922 CAS Number: 4775-98-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...