1,4-Bis(2-hydroxyethoxy)-2-butyne 70% (by GC)

Supplier: Ambeed
Warning

Synonyms: 1,4-Di(2-hydroxyethoxy)-2-butyne, 2,2-(But-2-yne-1,4-diylbis(oxy))diethanol, 2-Butyne-1,4-diol Bis(2-hydroxyethyl) Ether

A205689-1000G A205689-100G A205689-25G A205689-500G
77192-472EA 41.2 USD
77192-472 77192-474 77192-476 77192-478
1,4-Bis(2-hydroxyethoxy)-2-butyne 70% (by GC)
1,4-Bis(2-hydroxyethoxy)-2-butyne

Precaución: for Research Use Only

Formula: C₈H₁₄O₄
MW: 174.20 g/mol
MDL Number: MFCD00002883
CAS Number: 1606-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR