2-Fluorobenzonitrile 98%

Supplier: Ambeed
Warning

A205552-1000G A205552-100G A205552-10G A205552-25G A205552-500G A205552-5G
77164-750EA 116 USD
77164-750 77164-752 77164-754 77164-756 77164-758 77164-760
2-Fluorobenzonitrile 98%
2-Fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄FN
MDL Number: MFCD00001773
CAS Number: 394-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR