AZD-6738 98%

Supplier: Ambeed
Warning

A205112-10MG A205112-1MG A205112-2MG A205112-50MG A205112-100MG
77192-456EA 76 USD
77192-456 77192-458 77192-460 77615-726 77746-268
AZD-6738 98%
AZD-6738

Precaución: for Research Use Only

Formula: C₂₀H₂₄N₆O₂S
MDL Number: MFCD28952790
CAS Number: 1352226-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR