INCB-024360 ≥98%

Supplier: Ambeed
Warning

Synonyms: Epacadostat, (Z)-N-(3-Bromo-4-fluorophenyl)-N'-hydroxy-4-[2-(sulphamoylamino)ethylamino]-1,2,5-oxadiazole-3-carboxamidine

A205097-100MG A205097-10MG A205097-1G A205097-1MG A205097-250MG A205097-5MG A205097-2MG A205097-50MG
77164-738EA 109.18 USD
77164-738 77164-740 77164-742 77164-744 77164-746 77164-748 77612-004 77683-578
INCB-024360 ≥98%
INCB-024360

Precaución: For research use only.

Formula: C₁₁H₁₃BrFN₇O₄S
MW: 438.23 g/mol
MDL Number: MFCD26142934
CAS Number: 1204669-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR