Ibudilast 98%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Isopropylpyrazolo[1,5-a]pyridin-3-yl)-2-methylpropan-1-one, 3-Isobutyryl-2-isopropylpyrazolo[1,5-a]pyridine

A204764-100MG A204764-10MG A204764-50MG A204764-5MG
77192-438EA 25.75 USD
77192-438 77192-440 77192-442 77192-444
Ibudilast 98%
Ibudilast

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₂O
MW: 230.31 g/mol
MDL Number: MFCD00864808
CAS Number: 50847-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR