6-Bromo-4-chloro-2-methylbenzo[d]thiazole 95%

Supplier: Ambeed
Warning

A204116-100MG A204116-1G A204116-250MG
77164-698EA 64.89 USD
77164-698 77164-700 77164-702
6-Bromo-4-chloro-2-methylbenzo[d]thiazole 95%
6-Bromo-4-chloro-2-methylbenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₈H₅BrClNS
MW: 262.55 g/mol
MDL Number: MFCD23705860
CAS Number: 1427363-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR