2-Bromo-4-fluoro-1,3,5-trimethylbenzene 95%

Supplier: Ambeed
Warning

A204065-10G A204065-1G A204065-5G
77164-692EA 201.88 USD
77164-692 77164-694 77164-696
2-Bromo-4-fluoro-1,3,5-trimethylbenzene 95%
2-Bromo-4-fluoro-1,3,5-trimethylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrF
MW: 217.08 g/mol
MDL Number: MFCD22683870
CAS Number: 1580-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR