(3-Methoxyphenyl)methanesulfonamide 97%

Supplier: Ambeed
Warning

A203953-250MG A203953-5G A203953-1G
77164-686EA 27.81 USD
77164-686 77164-688 77164-690 77892-285
(3-Methoxyphenyl)methanesulfonamide 97%
(3-Methoxyphenyl)methanesulfonamide

Precaución: for Research Use Only

Formula: C₈H₁₁NO₃S
MDL Number: MFCD16671946
CAS Number: 89782-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR