Parecoxib 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: N-[[4-(5-Methyl-3-phenylisoxazol-4-yl)phenyl]sulphonyl]propionamide, N-((4-(5-Methyl-3-phenylisoxazol-4-yl)phenyl)sulphonyl)propionamide

A203658-1G A203658-250MG A203658-25MG A203658-50MG A203658-25G A203658-5MG
77164-674EA 33.99 USD
77164-674 77164-676 77164-678 77164-680 77683-569 77683-570
Parecoxib 98%, Ambeed.Inc
Parecoxib

Precaución: For research use only.

Formula: C₁₉H₁₈N₂O₄S
MW: 370.42 g/mol
MDL Number: MFCD25976408
CAS Number: 198470-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR