Methyl-1-methylpiperidine-3-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-methylpiperidine-3-carboxylate

A203621-10G A203621-1G A203621-5G
77192-382EA 24.72 USD
77192-382 77192-384 77192-386
Methyl-1-methylpiperidine-3-carboxylate 97%
Methyl-1-methylpiperidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₅NO₂
MW: 157.2100 g/mol
MDL Number: MFCD00128793
CAS Number: 1690-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR