2-Chloro-4,5-dimethoxypyrimidine 95%

Supplier: Ambeed
Warning

A203424-100MG A203424-1G A203424-250MG A203424-5G
77164-666EA 41 USD
77164-666 77164-668 77164-670 77164-672
2-Chloro-4,5-dimethoxypyrimidine 95%
2-Chloro-4,5-dimethoxypyrimidine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O₂
MW: 174.59 g/mol
MDL Number: MFCD21098377
CAS Number: 1333240-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR