2,7-Dimethyl-1H-indole 96%

Supplier: Ambeed
Warning

A202869-1G A202869-5G
77164-656EA 77 USD
77164-656 77892-279
2,7-Dimethyl-1H-indole 96%
2,7-Dimethyl-1H-indole

Precaución: for Research Use Only

Formula: C₁₀H₁₁N
MDL Number: MFCD00022715
CAS Number: 5621-13-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR