Uprosertib ≥98%

Supplier: Ambeed
Warning

Synonyms: (S)-N-(1-Amino-3-(3,4-difluorophenyl)propan-2-yl)-5-chloro-4-(4-chloro-1-methyl-1H-pyrazol-5-yl)furan-2-carboxamide, GSK2141795, GSK-2141795, GSK 2141795, N-[(2S)-1-amino-3-(3,4-difluorophenyl)propan-2-yl]-5-chloro-4-(4-chloro-2-methylpyrazol-3-yl)furan-2-carboxamide, N-((S)-1-amino-3-(3,4-difluorophenyl)propan-2-yl)-5-chloro-4-(4-chloro-1-methyl-1H-pyrazol-5-yl)furan-2-carboxamide, Uprosertib, GSK 795, GSK795

A202830-10MG A202830-1MG A202830-50MG A202830-5MG
77164-648EA 113.3 USD
77164-648 77164-650 77164-652 77164-654
Uprosertib ≥98%
Uprosertib

Precaución: for Research Use Only

Formula: C₁₈H₁₆Cl₂F₂N₄O₂
MW: 429.25 g/mol
MDL Number: MFCD28144686
CAS Number: 1047634-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR