4-(5-Amino-1,3-dioxo-1,3-dihydroisoindol-2-yl)butyric acid 97%

Supplier: Ambeed
Warning

A202761-10G A202761-1G A202761-250MG A202761-5G
77192-370EA 126 USD
77192-370 77192-372 77192-374 77192-376
4-(5-Amino-1,3-dioxo-1,3-dihydroisoindol-2-yl)butyric acid 97%
4-(5-Amino-1,3-dioxo-1,3-dihydroisoindol-2-yl)butyric acid

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O₄
MDL Number: MFCD01893052
CAS Number: 18595-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR