4-Chloro-2-(methylthio)pyrimidine 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
4-Chloro-2-methylthiopyrimidine
A202666-10G
A202666-25G
A202666-5G
A202666-100G
A202666-1G
77164-642EA
15.45
USD
77164-642
77164-644
77164-646
77611-962
77683-565
4-Chloro-2-(methylthio)pyrimidine 98%
4-Chloro-2-(methylthio)pyrimidine
Precaución: For research use only.
Formula:
C₅H₅ClN₂S MW: 160.63 g/mol |
MDL Number:
MFCD00006083 CAS Number: 49844-90-8 UN: 1760 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...