1-(4-Bromophenyl)pyrrole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(4-Bromophenyl)-1H-pyrrole
A201959-10G
A201959-250MG
A201959-25G
A201959-5G
A201959-1G
77192-358EA
185.4
USD
77192-358
77192-360
77192-362
77192-364
77746-260
1-(4-Bromophenyl)pyrrole 97%
1-(4-Bromophenyl)pyrrole
Precaución: for Research Use Only
Formula:
C₁₀H₈BrN MW: 222.09 g/mol |
MDL Number:
MFCD00128296 CAS Number: 5044-39-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...