b-Amino-3-methoxybenzenepropanol 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-(3-methoxyphenyl)propan-1-ol

A201900-100MG A201900-250MG
77164-616EA 30.9 USD
77164-616 77164-618
b-Amino-3-methoxybenzenepropanol 97%
b-Amino-3-methoxybenzenepropanol

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₂
MW: 181.232 g/mol
MDL Number: MFCD10034616
CAS Number: 344359-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR