2,6-Dichloro-5-(trifluoromethyl)nicotinonitrile 95%

Supplier: Ambeed
Warning

A201839-100MG A201839-1G
77164-612EA 225.57 USD
77164-612 77567-924
2,6-Dichloro-5-(trifluoromethyl)nicotinonitrile 95%
2,6-Dichloro-5-(trifluoromethyl)nicotinonitrile

Precaución: For research use only.

Formula: C₇HCl₂F₃N₂
MW: 241.00 g/mol
MDL Number: MFCD28099467
CAS Number: 1319804-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR