Ethyl piperidine-2-carboxylate hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A201722-100G
A201722-10G
A201722-25G
A201722-500G
A201722-5G
77192-338EA
34
USD
77192-338
77192-340
77192-342
77192-344
77192-346
Ethyl piperidine-2-carboxylate hydrochloride 98%
Ethyl piperidine-2-carboxylate hydrochloride
Precaución: for Research Use Only
Formula:
C₈H₁₆ClNO₂ |
MDL Number:
MFCD00134518 CAS Number: 77034-33-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...