N-Methyl-1-(3-nitrophenyl)methanamine 98%

Supplier: Ambeed
Warning

Synonyms: N-Methyl-N-(3-nitrobenzyl)amine

A201245-1G A201245-25G A201245-5G
77192-318EA 20.6 USD
77192-318 77192-320 77192-322
N-Methyl-1-(3-nitrophenyl)methanamine 98%
N-Methyl-1-(3-nitrophenyl)methanamine

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
MDL Number: MFCD04115408
CAS Number: 19499-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR