1-(4-Chlorophenyl)biguanide hydrochloride 95%

Supplier: Ambeed
Warning

A201185-100MG A201185-1MG
77192-308EA 13.39 USD
77192-308 77192-310
1-(4-Chlorophenyl)biguanide hydrochloride 95%
1-(4-Chlorophenyl)biguanide hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁Cl₂N₅
MW: 248.12 g/mol
MDL Number: MFCD00053020
CAS Number: 4022-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR