tert-Butyl (3-bromopyridin-4-yl)carbamate 97%

Supplier: Ambeed
Warning

A201055-10G A201055-1G A201055-5G A201055-250MG
77164-602EA 51 USD
77164-602 77164-604 77611-950 77892-275
tert-Butyl (3-bromopyridin-4-yl)carbamate 97%
tert-Butyl (3-bromopyridin-4-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrN₂O₂
MDL Number: MFCD01860257
CAS Number: 257937-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR