α,α,α,4-Tetrafluoro-o-toluic acid 98%

Supplier: Ambeed
Warning

A200691-10G A200691-1G A200691-25G A200691-5G A200691-100G
77192-286EA 13.39 USD
77192-286 77192-288 77192-290 77192-292 77550-284
α,α,α,4-Tetrafluoro-o-toluic acid 98%
α,α,α,4-Tetrafluoro-o-toluic acid

Precaución: for Research Use Only

Formula: C₈H₄F₄O₂
MW: 208.11 g/mol
MDL Number: MFCD00040982
CAS Number: 141179-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR