1,4-Bis(dimethylvinylsilyl)benzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1,4-Bis(dimethyl(vinyl)silyl)benzene, 1,4-Phenylenebis[dimethyl(vinyl)silane]
A200585-100G
A200585-10G
A200585-1G
A200585-25G
A200585-5G
77192-270EA
272.95
USD
77192-270
77192-272
77192-274
77192-276
77192-278
1,4-Bis(dimethylvinylsilyl)benzene 98%
1,4-Bis(dimethylvinylsilyl)benzene
Precaución: for Research Use Only
Formula:
C₁₄H₂₂Si₂ MW: 246.50 g/mol |
MDL Number:
MFCD07778038 CAS Number: 4519-17-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...