7-Chloro-1H-indazole 97%

Supplier: Ambeed
Warning

Synonyms: 7-Chloroindazole

A200575-1G A200575-250MG A200575-5G
77164-578EA 17.51 USD
77164-578 77164-580 77164-582
7-Chloro-1H-indazole 97%
7-Chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MW: 152.5810 g/mol
MDL Number: MFCD07368214
CAS Number: 37435-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR