Methyl-4-bromo-2,6-dimethylbenzoate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4-bromo-2,6-dimethylbenzoate
A200361-10G
A200361-1G
A200361-250MG
A200361-25G
A200361-5G
77164-550EA
273.98
USD
77164-550
77164-552
77164-554
77164-556
77164-558
Methyl-4-bromo-2,6-dimethylbenzoate 95%
Methyl-4-bromo-2,6-dimethylbenzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₁BrO₂ MW: 243.11 g/mol |
MDL Number:
MFCD11227152 CAS Number: 90841-46-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...