Methyl-3-amino-5-bromopicolinate 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-5-bromopicolinate

A200342-1G A200342-250MG A200342-5G A200342-25G
77164-544EA 17 USD
77164-544 77164-546 77164-548 77683-558
Methyl-3-amino-5-bromopicolinate 98%, Ambeed.Inc
Methyl-3-amino-5-bromopicolinate

Precaución: For research use only.

Formula: C₇H₇BrN₂O₂
MW: 231.06 g/mol
MDL Number: MFCD17015929
CAS Number: 1072448-08-8
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR