1,1-Dibromopinacolone 98%

Supplier: Ambeed
Warning

A199855-1G A199855-25G A199855-5G
77192-244EA 14 USD
77192-244 77192-246 77192-248
1,1-Dibromopinacolone 98%
1,1-Dibromopinacolone

Precaución: for Research Use Only

Formula: C₆H₁₀Br₂O
MDL Number: MFCD00045030
CAS Number: 30263-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR