3',5'-Dichloro-2,2,2,4'-tetrafluoroacetophenone 95%

Supplier: Ambeed

A198810-100G A198810-10G A198810-1G A198810-25G A198810-5G
77164-510EA 150 USD
77164-510 77164-512 77164-514 77164-516 77164-518
3',5'-Dichloro-2,2,2,4'-tetrafluoroacetophenone 95%
3',5'-Dichloro-2,2,2,4'-tetrafluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₂Cl₂F₄O
MDL Number: MFCD23382507
CAS Number: 1190865-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR