5,6-Dihydroxypyrimidine-4-carboxylic acid 96%

Supplier: Ambeed
Warning

A198497-100MG A198497-1G A198497-250MG
77164-504EA 177.16 USD
77164-504 77164-506 77164-508
5,6-Dihydroxypyrimidine-4-carboxylic acid 96%
5,6-Dihydroxypyrimidine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₄N₂O₄
MW: 156.09 g/mol
MDL Number: MFCD09701387
CAS Number: 52412-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR