3-Bromo-5-hydroxybenzonitrile 98%

Supplier: Ambeed
Warning

A198034-100MG A198034-250MG A198034-1G
77164-498EA 24.72 USD
77164-498 77164-500 77567-860
3-Bromo-5-hydroxybenzonitrile 98%
3-Bromo-5-hydroxybenzonitrile

Precaución: For research use only.

Formula: C₇H₄BrNO
MW: 198.0200 g/mol
MDL Number: MFCD16659096
CAS Number: 770718-92-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR