1-Acetylnaphthalene 97%

Supplier: Ambeed
Warning

A197827-1000G A197827-100G A197827-10G A197827-25G A197827-500G
77192-198EA 66 USD
77192-198 77192-200 77192-202 77192-204 77192-206
1-Acetylnaphthalene 97%
1-Acetylnaphthalene

Precaución: for Research Use Only

Formula: C₁₂H₁₀O
MDL Number: MFCD00004013
CAS Number: 941-98-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR