Pyridoxal-5'-phosphate monohydrate 97%

Supplier: Ambeed
Warning

Synonyms: Pyridoxal-5-phosphate hydrate, Pyridoxal 5-phosphate monohydrate, Pyridoxal 5'-phosphate monohydrate, (4-Formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate hydrate

A197820-25G A197820-5G A197820-100G A197820-1G A197820-500G
77192-194EA 41 USD
77192-194 77192-196 77567-852 77593-278 77746-247
Pyridoxal-5'-phosphate monohydrate 97%
Pyridoxal 5'-phosphate monohydrate

Precaución: For research use only.

Formula: C₈H₁₀NO₆P·1H₂O
MW: 265.17 g/mol (247.15 anhy)
MDL Number: MFCD00149414
CAS Number: 41468-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR